Capillary columns, Elite-225 for autosystem
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N9316175 N9316177 N9316181
PERCN9316175EA 580 EUR
PERCN9316175 PERCN9316177 PERCN9316181
Capillary columns, Elite-225 for autosystem
Kolommen voor chromatografie
Elite-225 columns are fused silica columns coated with phenylmethylpolysiloxane (50% cyanopropylphenyl).