Test tubes with standard ground joints
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2.580.125.BR 2.582.510 2.582.550
WITG2.580.125.BREA 10.4 EUR
WITG2.580.125.BR WITG2.582.510 WITG2.582.550
Test tubes with standard ground joints
Buisjes Reageerbuisjes
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Round bottom, graduated or ungraduated. With standard ground joints and PE stopper.