Volumetric flasks, class A, white markings
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3.670.012 3.670.500 3.670.00114 3.670.030 3.670.150 3.670.017 3.670.018 3.670.016 3.670.025.29 3.669.001.10BR
WITG3.670.012EA 42.9 EUR
WITG3.670.012 WITG3.670.500 WITG3.670.00114 WITG3.670.030 WITG3.670.150 WITG3.670.017 WITG3.670.018 WITG3.670.016 WITG3.670.025.29 WITG3.669.001.10BR
Volumetric flasks, class A, white markings
Kolven Maatkolven
Borosilicate glass 3.3, amber, with glass stopper.
  • White markings
  • Conformity certified (DE-M marking)
  • Inclusive batch certificate
Certificaten: DIN EN ISO 1042