Measuring cylinders, tall form, class B, borosilicate glass
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3.700.200 3.700.003
613-5248EA 86.9 EUR
613-5248 WITG3.700.003
Measuring cylinders, tall form, class B, borosilicate glass
Cilinders Maatcilinders
Borosilicate glass 3.3, tall form, class B.
  • Calibrated to contain (TC, In)
  • Hexagonal with plastic base and protection collar, spout
  • Brown graduation
Certificaten: DIN EN ISO 4788