Centrifuge tubes, round bottom
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4.940.105 4.940.002 4.940.005 4.945.104 4.945.103 4.940.103
WITG4.940.105EA 20.1 EUR
WITG4.940.105 WITG4.940.002 WITG4.940.005 WITG4.945.104 WITG4.945.103 WITG4.940.103
Centrifuge tubes, round bottom
Buisjes Centrifugebuisjes
DURAN®, borosilicate glass.
  • Heavy wall
  • Very good chemical resistance
  • Very good temperature resistance
Round bottom, rimless.