Desiccators with PROTEFAN safety coating
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5.065.200 5.065.300
WITG5.065.200EA 438 EUR
WITG5.065.200 WITG5.065.300
Desiccators with PROTEFAN safety coating
Exsiccatoren Exsiccatoren, glas
Desiccators are made of borosilicate glass, with safety PROTEFAN coating as implosion and splinter protection.