Space tubing, Infusomat®
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8700036SP 8700095SP 8250731SP
BRAU8700036SPEA 1300 EUR
BRAU8700036SP BRAU8700095SP BRAU8250731SP
Space tubing, Infusomat®
Infusen
Convenient drip chamber design with sharp spike ensures easy piercing of container ports, bacteria proof venting (closable), 20 drops 1 ml ±0,1 ml (aqua dest.) and fluid filter.
  • Silicone pump segment for high delivery accuracy and long-term consistency
  • Roller clamp with line holder and unique spike protector for safe disposal
  • Neutrapur® versions are PVC-free
Different fastening clips on the lower and upper part of the silicone pump segment for easy and secure tubing guidance. Additional set based anti-free flow clamp, automatically clamps the lineMore Product Information