Trypsinising flasks
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5.507.305 5.507.308 5.507.306 5.507.303 5.507.304 5.507.307
WITG5.507.305EA 58.7 EUR
WITG5.507.305 WITG5.507.308 WITG5.507.306 WITG5.507.303 WITG5.507.304 WITG5.507.307
Trypsinising flasks
Kolven Trypsinisatie kolven
Trypsinising flasks, graduated, designed for converting tissue fragments and homogenates into cell suspensions by trypsin digestion of connective proteins. Flutes in flask enhance vigorous agitation.