Volumetric flasks, class A, amber markings
Leverancier: Poulten & Graf
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.50037F 1.50043F 1.50045F 1.50051F 1.50053F 1.50055F 1.50059F
POUL1.50037FEA 12.3 EUR
POUL1.50037F POUL1.50043F 612-0550 POUL1.50051F POUL1.50053F 612-0533 POUL1.50059F
Volumetric flasks, class A, amber markings
Kolven Maatkolven
Borosilicate glass 3.3 with beaded rim.
  • Calibrated to contain (TC, In)
  • With ground ring mark
  • Conformity certified (DE-M marking)
Certificaten: DIN EN ISO 1042