Analogue stopwatch, Allsport
Leverancier: Hanhart
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HANH122.6401-00EA 414 EUR
HANH122.6401-00
Analogue stopwatch, Allsport
Timers Analoge stopwatches
Universal referee’s stopwatch/addition timer in chromium plated brass case.
  • Pin lever, 7 jewels
  • Scale goes up to 60 minutes
  • 20, 35, 45 and 60 minutes marked in yellow
  • Shock-, dust- and waterproof