SteriWare® Liquid Funnels
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8370H-200S 8370H-100 8370B-100 8370B-100S 8370H-200 8370H-50 8370H-50S 8370H-100S
SAMP8370H-200SEA 184 EUR
SAMP8370H-200S 300-0592 300-0589 SAMP8370B-100S SAMP8370H-200 300-0590 300-0591 SAMP8370H-100S
SteriWare® Liquid Funnels
Trechters Filtertrechters Filtertrechters
Single use, disposable liquid funnels ideal for laboratories and manufacturing areas. Designed to use straight from bags.
  • Made of HDPE
  • Moulded and packed in a class 100,000 cleanroom
  • With small hygenic side handle
  • BSE/TSE free
  • Available sterile (gamma irradiated)
Certificaten: Material conforms to FDA, EU 10/2011 and EC 1935/2004. Full batch traceability.
Verpakking: Individually bagged.