Digital desktop timers, Prisma 200 / Prisma 400
Leverancier: HANHART
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
626.2625-00 625.2625-00
609-1114EA 216 EUR
609-1114 609-1113
Digital desktop timers, Prisma 200 / Prisma 400
Timers Digitale timers
Timers with two (Prisma 200) or four channels (Prisma 400).
  • Channels can be started simultaneously
  • Addition, Flyback, Countdown
  • Adjustable acoustic alarm, clock
  • Repeat, manual or automatic
  • Selectable: 1/10 second or 1/100 minute
  • Timing capacity: 10 hours