Stirring shafts, retreat curve
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005.80850.300 005.80850.400 005.80875.300 005.80875.400 005.81050.400 005.81050.500 005.81075.400 005.81075.500
COWI005.80850.300EA 78.4 EUR
COWI005.80850.300 COWI005.80850.400 COWI005.80875.300 COWI005.80875.400 COWI005.81050.400 COWI005.81050.500 COWI005.81075.400 COWI005.81075.500
Stirring shafts, retreat curve
Roerders Roerassen
Stirring shafts with a retreat angle of 30°.