Shelving systems, stem castor carts with solid bottom shelves, MetroMax i®
Leverancier: Metro International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5X567EGX3 5Q557EG3
METO5X567EGX3EA 2692 EUR
METO5X567EGX3 METO5Q557EG3
Shelving systems, stem castor carts with solid bottom shelves, MetroMax i®
Meubilair Legborden
Corrosion proof shelves and posts with removable polymer mats.
  • Microban® antimicrobial protection
  • Load capacity: 408 kg (for models with PU castors)
Leveringsinformatie: Standard unit consist of 5 shelves (4 open grid, 1 bottom solid shelf), posts, wedge connectors and polyurethane castors.