Sampling spoons, sterile
Leverancier: Nasco
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NASCB01041AWAEA 66.4 EUR
NASCB01041AWA
Sampling spoons, sterile
Lepels
PS, white. These gas sterilised spoons are ideal for collecting samples.
  • Made of sturdy, durable PS
  • Ideal for use with Whirl-Pak® bags
The procedure for contamination-free use is to open the bag, push one spoon up, pull it out, and reclose the bag.
Verpakking: 20 per bag, 100 per box.