RevertAid™ Reverse Transcriptase
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EP0441 EP0442
EP0441EA 91.5 EUR
EP0441 EP0442
RevertAid™ Reverse Transcriptase
Reagentia voor nucleïnezuren RT-PCR and cDNA Reagents
RevertAid™ Reverse Transcriptase (RT) is a genetically modified M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity and a RNase H activity specific to RNA in RNA-DNA hybrids, which is significantly lower than that of Avian Myeloblastosis Virus (AMV) reverse transcriptase.
  • Efficient synthesis of full length first strand cDNA up to 13 kb
  • Optimum activity at 42 °C
  • Active up to 50 °C
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labelled nucleotides)
Leveringsinformatie: Supplied with 5X reaction buffer.