Wire baskets, square
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41075016 41075020
HECH41075016EA 127 EUR
HECH41075016 HECH41075020
Wire baskets, square
Manden Draadmanden
These square baskets are made of stainless steel. Designed for transporting, storing, cleaning and sterilising laboratory utensils.
  • Excellent temperature resistance
  • Good chemical resistance
  • Reinforced edges