PCR master mix (2X)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
K0171
K0172
K0171EA
132
EUR
K0171
K0172
PCR master mix (2X)
Reagentia voor nucleïnezuren
End-point PCR Enzymes and Kits
PCR Master Mix is a 2X concentrated solution of Taq DNA polymerase, dNTPs and all other components required for PCR, except DNA template and primers. This pre-mixed formulation saves time and reduces contamination due to a reduced number of pipetting steps required for PCR set up. The mix is optimised for efficient and reproducible PCR.