RevertAid™ H minus first strand cDNA synthesis kits
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
K1631 K1632
K1631EA 135 EUR
K1631 K1632
RevertAid™ H minus first strand cDNA synthesis kits
Reagentia voor nucleïnezuren RT-PCR and cDNA Reagents
RevertAid™ H Minus First Strand cDNA Synthesis Kit is a complete system for efficient synthesis of first strand cDNA from RNA templates. The kit includes RevertAid™ H Minus Reverse Transcriptase, which has a point mutation that completely eliminates RNase H activity. Therefore, it does not degrade RNA in RNA-DNA hybrids during synthesis of the first strand cDNA.
  • High yields of full-length first strand cDNA up to 13 kb
  • Increased reaction temperatures in the range of 42 to 55 °C
  • Supplied with the recombinant RiboLock™ RNase Inhibitor
  • Complete kit – oligo(dT)₁₈ and random hexamer primers included
Leveringsinformatie: Each kit includes RevertAid™ H Minus Reverse Transcriptase, RiboLock™ RNase Inhibitor, 5X Reaction Buffer, dNTP Mix, Oligo(dT)₁₅ Primer, Random Hexamer Primer, Control RNA, Control Primers and nuclease-free water.