Utility Cart
Leverancier: Rubbermaid Commercial Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FG342488OWHT FG342488BLA FG342488PLAT
129-0448EA 551 EUR
129-0448 129-0424 129-0427
Utility Cart
Transportwagens
Trolleys are made of PP with four swivel castors.
  • Easy to clean, smooth surfaces
  • Plastic construction help to reduce noise
  • Ergonomic, easy grip handles
  • Quiet and large 102 mm non marking swivel castors
Load capacity: 90,7 kg