(2,4-Difluorophenyl)acetonitrile
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2,4-Difluorobenzyl cyanide, 2-(2,4-Difluorophenyl)acetonitrile
HAN00045.1 HAN00045.10
MAYBHAN00045.1EA 0 EUR
MAYBHAN00045.1 MAYBHAN00045.10
(2,4-Difluorophenyl)acetonitrile
(2,4-Difluorophenyl)acetonitrile
Formule: C₈H₅F₂N
Kookpunt: 90 °C (0,3 mmHg)
Dichtheid: 1,2503 g/cm³ (15 °C)
MDL Nummer: MFCD00009971
CAS nummer: 656-35-9
EINECS: 211-514-1
UN: 3276
ADR: 6.1,III