Chenodeoxycholic acid
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 3α,7α-Dihydroxycholanic acid, 7α-Hydroxylithocholic acid, 7alpha-Hydroxylithocholic acid, 3α,7α-Dihydroxy-5β-cholan-24-oic acid, 3α,7α-Dihydroxycholan-24-oic acid, Chenodiol, 3α,7α-Dihydroxy-5β-cholanic acid
10011286-1 10011286-5 10011286-10 10011286-25
CAYM10011286-1EA 49.2 EUR
CAYM10011286-1 CAYM10011286-5 CAYM10011286-10 CAYM10011286-25
Chenodeoxycholic acid
Chenodeoxycholic acid
Formule: C₂₄H₄₀O₄
MW: 392.58 g/mol
Smeltpunt: 164.85…165.85 °C
Dichtheid: 1.162 g/cm³ (20 °C)
MDL Nummer: MFCD00064142
CAS nummer: 474-25-9
EINECS: 207-481-8
Merck Index: 12,02096