12-Tricosanone ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

T0404-25G T0404-500G
TCIAT0404-25GEA 74.8 EUR
TCIAT0404-25G TCIAT0404-500G
12-Tricosanone ≥95.0% (by GC)
12-Tricosanone
Formule: C₂₃H₄₆O
MW: 338.62 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00026562
CAS nummer: 540-09-0
EINECS: 208-735-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdbbd92a500143a137658ca8ec03654003920444&vid=46ced39eaa0bc637dca749c83317d4c594615e10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Melting point 68.0 to 72.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR