4-Chlorophenylsulphonylacetone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-(4-Chlorophenylsulphonyl)acetone, 4-Chlorobenzenesulphonylacetone

KM01166.1 KM01166.10 KM01166.50
MAYBKM01166.1EA 0 EUR
MAYBKM01166.1 MAYBKM01166.10 MAYBKM01166.50
4-Chlorophenylsulphonylacetone
4-Chlorophenylsulphonylacetone
Formule: C₉H₉ClO₃S
MW: 232.69 g/mol
MDL Nummer: MFCD00018663
CAS nummer: 5000-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR