3-Bromothiophenol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B2348-25G
B2348-5G
TCIAB2348-25GEA
234
EUR
TCIAB2348-25G
TCIAB2348-5G
3-Bromothiophenol ≥98.0% (by GC)
3-Bromothiophenol
Formule:
C₆H₅BrS Kookpunt: 121°C Dichtheid: 1,583 g/cm³ (30 °C) Vlampunt: 105°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00009603 CAS nummer: 6320-01-0 EINECS: 228-664-9 UN: 1760 ADR: 8,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...