(R)-(+)-2,3-Epoxy-1-propanol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
(R)-(+)-2-(Hydroxymethyl)oxirane, (R)-(+)-1-Hydroxy-2,3-epoxypropane, (R)-Oxiran-2-ylmethanol, (R)-(+)-Glycidol
G0363-25G
G0363-5G
TCIAG0363-25GEA
279
EUR
TCIAG0363-25G
TCIAG0363-5G
(R)-(+)-2,3-Epoxy-1-propanol ≥98.0% (by GC)
(R)-(+)-2,3-Epoxy-1-propanol
Formule:
C₃H₆O₂ Kookpunt: 57°C Vlampunt: 72°C Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00074873 CAS nummer: 57044-25-4 EINECS: 404-660-4 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Specific rotation [a]20/D | +13.0 to +17.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...