3-Acetylcoumarin

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB03234DA BTB03234EA BTB03234.50
MAYBBTB03234DAEA 0 EUR
MAYBBTB03234DA MAYBBTB03234EA MAYBBTB03234.50
3-Acetylcoumarin
3-Acetylcoumarin
Formule: C₁₁H₈O₃
MDL Nummer: MFCD00006853
CAS nummer: 3949-36-8
EINECS: 223-541-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR