Diphenylacetyl chloride 97.5%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
292020050 292020250
ACRO292020050EA 44.8 EUR
ACRO292020050 ACRO292020250
Diphenylacetyl chloride 97.5%
Diphenylacetyl chloride
Formule: C₁₄H₁₁ClO
Kookpunt: 175…176 °C (17 mmHg)
Smeltpunt: 51…56 °C
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013655
CAS nummer: 1871-76-7
EINECS: 217-493-5
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 51 to 56 °C
GC ≥96.5 %