Thulium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
11200.04
11200.09
11200.04EA
130
EUR
11200.04
11200.09
Thulium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Thulium(III) chloride hydrate
Purity: 99.9%
CAS No.: 19423-86-0
Molecular Formula: Cl3Tm.xH2O
Formula Weight: 275.29(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Solubility: Soluble in water, alcohol
Merck Reference: 14,9394
MDL No.: MFCD00011283
Formule:
Cl₃Tm·H₂O MW: 293.309 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149877 CAS nummer: 19423-86-0 EINECS: 236-904-9 Merck Index: 14,9394 |