2-Chloro-3-methylquinoline
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NRB00907DA
NRB00907EA
MAYBNRB00907DAEA
0
EUR
MAYBNRB00907DA
MAYBNRB00907EA
2-Chloro-3-methylquinoline
2-Chloro-3-methylquinoline
Formule:
C₁₀H₈ClN MW: 177.63 g/mol |
MDL Nummer:
MFCD00100265 CAS nummer: 57876-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...