2,2-Diphenylacetamide ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
L08485.06
L08485.14
L08485.06EA
69.2
EUR
L08485.06
L08485.14
2,2-Diphenylacetamide ≥98%
2,2-Diphenylacetamide
Purity: 98%
CAS No.: 4695-13-0
Molecular Formula: C14H13NO
Formula Weight: 211.26
Melting Point: 164-167°
Beilstein Reference: 1871583
Merck Reference: 14,3316
MDL No.: MFCD00025496
Formule:
C₁₄H₁₃NO Smeltpunt: 164…167 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00025496 CAS nummer: 4695-13-0 Merck Index: 14,3316 |