5-Fluoro-2'-deoxyuridine (floxuridine) ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine
L16497.MC L16497.ME
L16497.MCEA 56.2 EUR
L16497.MC L16497.ME
5-Fluoro-2'-deoxyuridine (floxuridine) ≥98%
5-Fluoro-2'-deoxyuridine (floxuridine)
Purity: 98+% CAS No.: 50-91-9 Molecular Formula: C9H11FN2O5 Formula Weight: 246.20 Melting Point: 148-151° Beilstein Reference: 2645818 Merck Reference: 14,4112 UN No.: 2811 MDL No.: MFCD00006530
Formule: C₉H₁₁FN₂O₅
MW: 246.19 g/mol
Smeltpunt: 148…151 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006530
CAS nummer: 50-91-9
EINECS: 200-072-5
UN: 2811
ADR: 6.1 III