Diethyl (cyanomethyl)phosphonate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01599.1 SB01599.10 SB01599.50
MAYBSB01599.1EA 0 EUR
MAYBSB01599.1 MAYBSB01599.10 MAYBSB01599.50
Diethyl (cyanomethyl)phosphonate
Diethyl (cyanomethyl)phosphonate
Formule: C₆H₁₂NO₃P
MDL Nummer: MFCD00001893
CAS nummer: 2537-48-6
EINECS: 219-806-0
UN: 3278
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR