2,3-Dihydrobenzo[b]furan-7-carboxylic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D2446-1G D2446-5G
TCIAD2446-1GEA 95.7 EUR
TCIAD2446-1G TCIAD2446-5G
2,3-Dihydrobenzo[b]furan-7-carboxylic acid ≥98.0% (by titrimetric analysis)
2,3-Dihydrobenzo[b]furan-7-carboxylic acid
Formule: C₉H₈O₃
MDL Nummer: MFCD00671526
CAS nummer: 35700-40-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f83570da6ec36ddb99b3a9de0132a5a9b5618f35&vid=f83570da6ec36ddb99b3a9de0132a5a9b5618f35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 171.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR