Thionin(acetate), Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 3,7-Diaminophenothiazin-5-ium acetate
MQ200
861340-25G 861340-5G
861340-25GEA 487 EUR
861340-25G 861340-5G
Thionin(acetate), Sigma-Aldrich®
Thionin(acetate)
Formule: C₁₄H₁₃N₃O₂S
MW: 287.34 g/mol
MDL Nummer: MFCD00081194
CAS nummer: 78338-22-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e52365a2e45e482c74604f1f41c182c4bdb558aa&vid=2424442daa6961cb6e6ab89cb4d2222b1e376bca&prid=web&clid=SAPDAM