L-Methionine methyl ester hydrochloride
Leverancier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4000448.0025
4000448.0100
BACE4000448.0025EA
0
EUR
BACE4000448.0025
BACE4000448.0100
L-Methionine methyl ester hydrochloride
L-Methionine methyl ester hydrochloride
Formule:
C₆H₁₃NO₂S·HCl |
MDL Nummer:
MFCD00012491 CAS nummer: 2491-18-1 EINECS: 219-651-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...