5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
A155.1 A155.2 A155.3 A155.4
ROTHA155.1EA 34.9 EUR
ROTHA155.1 ROTHA155.2 ROTHA155.3 ROTHA155.4
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Formule: C₈H₄BrClNNa₂O₄P
MW: 370.43 g/mol
MDL Nummer: MFCD00036757
CAS nummer: 102185-33-1