5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: BCIP, P-toluidine 5-bromo-4-chloro-1H-indol-3-yl phosphate
A1117.0001 A1117.0005
A1117.0001EA 301 EUR
A1117.0001 A1117.0005
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Formule: C₁₅H₁₅BrClN₂O₄P
MW: 433.6214 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00040636
CAS nummer: 6578-06-9
EINECS: 229-506-1