Boron trifluoride acetic acid complex (1:2) (35 - 36,5% BF₃)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Boron fluoride, acetic acid complex, Boron trifluoride - acetic acid complex, Trifluoroborane compound with acetic acid (1:2)
A11471.AU A11471.AK
A11471.AUEA 177 EUR
A11471.AU A11471.AK
Boron trifluoride acetic acid complex (1:2) (35 - 36,5% BF₃)
Boron trifluoride acetic acid complex (1:2)
Purity: 96% CAS No.: 373-61-5 Molecular Formula: BF3.2CH3CO2H Formula Weight: 187.91 Flash Point: 83°(181°F) Refractive Index: 1.3670 Odour: Pungent pH: 1 Melting Point: -47° Boiling Point: 143-146° Beilstein Reference: 3686177 UN No.: 1742 MDL No.: MFCD00036359
Formule: C₄H₈BF₃O₄
MW: 187.91 g/mol
Kookpunt: 143…146 °C
Smeltpunt: –47 °C
Dichtheid: 1.353
Vlampunt: 83 °C (181 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036359
CAS nummer: 373-61-5
EINECS: 206-768-5
UN: 1742
ADR: 8 II