Indomethacin 98
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A19910.14
A19910.06
A19910.22
A19910.14EA
168
EUR
A19910.14
A19910.06
A19910.22
Indomethacin 98
Indomethacin
Purity: 98%
CAS No.: 53-86-1
Molecular Formula: C19H16ClNO4
Formula Weight: 357.8
Melting Point: 158-161°
Beilstein Reference: 497341
Merck Reference: 14,4968
UN No.: 2811
MDL No.: MFCD00057095
Formule:
C₁₉H₁₆ClNO₄ MW: 357.80 g/mol Smeltpunt: 158…161 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00057095 CAS nummer: 53-86-1 EINECS: 200-186-5 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...