Luminol (3-Aminophthalhydrazide)
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: NSC 5064
4203.1 4203.2
ROTH4203.1EA 0 EUR
ROTH4203.1 ROTH4203.2
Luminol (3-Aminophthalhydrazide)
Luminol (3-Aminophthalhydrazide)
Formule: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Nummer: MFCD00006890
CAS nummer: 521-31-3
EINECS: 208-309-4