Luminol (3-Aminophthalhydrazide)

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

28085.02 28085.03
SERA28085.02EA 44 EUR
SERA28085.02 SERA28085.03
Luminol (3-Aminophthalhydrazide)
Luminol (3-Aminophthalhydrazide)
Formule: C₈H₇N₃O₂
MDL Nummer: MFCD00006890
CAS nummer: 521-31-3
EINECS: 208-309-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR