Magnetic stirring bars, glass-covered
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001.1219 001.1250
COWI001.1219EA 56.2 EUR
COWI001.1219 COWI001.1250
Magnetic stirring bars, glass-covered
Roerders Magnetische roerstaven
Magnetic stirring bars designed for use with very abrasive media which may erode PTFE.
  • Cylindrical in shape with Alnico V magnetic core