Measuring cylinders, tall form, class A, borosilicate glass, with USP certificate
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
224015427 224016027 224017027 224017527 224018027 224018627 224019027 224019327 224019427
HIRS224015427EA 14.1 EUR
HIRS224015427 HIRS224016027 HIRS224017027 HIRS224017527 HIRS224018027 HIRS224018627 HIRS224019027 HIRS224019327 HIRS224019427
Measuring cylinders, tall form, class A, borosilicate glass, with USP certificate
Cilinders Maatcilinders
DURAN® Borosilicate glass, Class A.
  • Blue graduation, main point ring graduation
  • Hexagonal glass base, spout
  • With dated batch identification
USP certificate downloadable.