PIG® absorbent pillows
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PIL201 PIL204 PIL205
121-0539EA 139 EUR
121-0539 NEWPPIL204 NEWPPIL205
PIG® absorbent pillows
Absorptiematerialen en bescherming tegen morsen
Add extra absorbency to your spill cleanup efforts with high-capacity PIG Pillows. Also great for steady leaks and drips.
  • Pillow has large surface area, high capacity and fast-wicking filler to quickly soak up liquids
  • Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated
  • Cellulose filler is highly absorbent for long-term use or big spill cleanup
  • Compact design is great for catching persistent drips and leaks in small spaces or adding absorbency for spill response
  • Absorbs and retains oils, coolants, solvents and water; not recommended for acids, caustics or other corrosive liquids
85% mixture of pre- and post-consumer recycled cellulose filler. Can be incinerated after use to reduce waste or for fuels blending.