Membrane dialysis discs and sheets, Spectra/Por®
Leverancier: Spectrum Laboratories
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
132476 132478 132480 132482 132486 132488 132677 132686 132723
SPEC132476EA 264 EUR
SPEC132476 SPEC132478 SPEC132480 SPEC132482 SPEC132486 SPEC132488 SPEC132677 SPEC132686 SPEC132723
Membrane dialysis discs and sheets, Spectra/Por®
Filters Membraanfilters
Pre-cut membrane discs are used with Spectra/Por® MacroDialyzers and Equilibrium Dialyzers.
  • Cut square membrane sheets can be used for a variety of dialysis equipment