Lids for reaction vessels
Leverancier: KGW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D4278-S D4303-S D4304
KGWAD4278-SEA 0 EUR
KGWAD4278-S KGWAD4303-S KGWAD4304
Lids for reaction vessels
Deksels Deksels voor reactievaten
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Order Now
Specificaties
Environmentally Preferable