Cans, waste disposal
Leverancier: KGW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1621 1622 1625
119-1008EA 918 EUR
119-1008 119-1015 119-1004
Cans, waste disposal
Veiligheidscontainers
Insert made of borosilicate glass 3.3. Ideal for storage and transport of aggressive waste in laboratories.
  • Good tipping stability
  • Glass is resistant to all kind of chemicals
  • Very good temperature resistance
Certificaten: In accordance with DIN 12111, DIN 12116, DIN 52322.
Order Now
Specificaties
Environmentally Preferable