Cork borer sets
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41593006 41593012
HECH41593006EA 63.1 EUR
HECH41593006 HECH41593012
Cork borer sets
Kurkboren
These cork borers are made of nickel-plated brass tubes with handles.