Funnels, powder
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5.790.004 5.790.001
WITG5.790.004EA 18.2 EUR
WITG5.790.004 WITG5.790.001
Funnels, powder
Trechters Poedertrechters
Powder funnels, made of DURAN® borosilicate glass 3.3, with short wide stem.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Order Now
Specificaties
Environmentally Preferable